Gene Transcripts
- Gene Symbol
- Gm29680
- Entrez Gene ID
- 544960
- Gene Name
- predicted gene, 29680
- Gene Aliases
- Chromosome Location
- Chr. 13: 90179217 - 90217142 on Build GRCm38
- UniGene ID
- Mm.316393
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK144747.1 |
- |
6 |
2548 |
|
View Full Product Details