Gene Transcripts
- Gene Symbol
- Gm15396
- Entrez Gene ID
- 102632404
- Gene Name
- predicted gene 15396
- Gene Aliases
- OTTMUSG00000021118
- Chromosome Location
- Chr. 7: 44580300 - 44590619 on Build GRCm38
- UniGene ID
- Mm.383181
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131623.1 |
- |
2 |
273 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ041946.1 |
- |
2 |
273 |
|
Chr. 7 - 44580353 - 44590624 [-] |
View Full Product Details