Gene Transcripts
- Gene Symbol
- 4930452N14Rik
- Entrez Gene ID
- 78112
- Gene Name
- RIKEN cDNA 4930452N14 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 154241521 - 154256906 on Build GRCm38
- UniGene ID
- Mm.401935
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040629.1 |
|
1 |
101 |
|
GenBank mRNA |
AK019628.1 |
- |
1 |
101 |
|
View Full Product Details