Gene Transcripts
- Gene Symbol
- 4930520M14Rik
- Entrez Gene ID
- 78182
- Gene Name
- RIKEN cDNA 4930520M14 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019689.1 |
- |
1 |
122 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715405.1 |
- |
1 |
122 |
|
Chr. 5 - 139215495 - 139216356 [-] |
View Full Product Details