Gene Transcripts
- Gene Symbol
- Gm32496
- Entrez Gene ID
- 102635065
- Gene Name
- predicted gene, 32496
- Gene Aliases
- Chromosome Location
- Chr. 3: 8628051 - 8632454 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK143350.1 |
- |
1 |
116 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ066986.1 |
- |
1 |
116 |
|
Chr. 3 - 8628051 - 8632174 [+] |
View Full Product Details