Gene Transcripts
- Gene Symbol
- 4933430M04Rik
- Entrez Gene ID
- 71316
- Gene Name
- RIKEN cDNA 4933430M04 gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 24481909 - 24569951 on Build GRCm38
- UniGene ID
- Mm.79162
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045857.2 |
|
1 |
27 |
|
GenBank mRNA |
AK016999.1 |
- |
1 |
28 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716684.1 |
- |
1 |
28 |
|
Chr. 11 - 24481909 - 24569952 [+] |
View Full Product Details