Gene Transcripts
- Gene Symbol
- Ly6k
- Entrez Gene ID
- 76486
- Gene Name
- lymphocyte antigen 6 complex, locus K
- Gene Aliases
- 2410015A16Rik, 3110035B01Rik, mLy-6K
- Chromosome Location
- Chr. 15: 74796874 - 74799968 on Build GRCm38
- UniGene ID
- Mm.273319
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014121.1 |
- |
|
152 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY713107.1 |
- |
1 |
152 |
|
Chr. 15 - 74784065 - 74799966 [+] |
View Full Product Details