Gene Transcripts
- Gene Symbol
- 4930548K13Rik
- Entrez Gene ID
- 75360
- Gene Name
- RIKEN cDNA 4930548K13 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 26635820 - 26705449 on Build GRCm38
- UniGene ID
- Mm.67233
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040656.1 |
|
2 |
214 |
|
GenBank mRNA |
AK016071.1 |
- |
2 |
214 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN842203.1 |
- |
|
192 |
|
Chr. - - [-] |
GenBankmRNA |
BY715669.1 |
- |
2 |
214 |
|
Chr. 4 - 26635820 - 26705449 [-] |
View Full Product Details