Gene Transcripts
- Gene Symbol
- LOC105372582
- Entrez Gene ID
- 105372582
- Gene Name
- uncharacterized LOC105372582
- Gene Aliases
- Chromosome Location
- Chr. 20: 25751208 - 25756050 on Build GRCh38
- UniGene ID
- Hs.516951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135006.1 |
|
1, 2 |
130 |
|
GenBank mRNA |
DB215985.1 |
- |
1, 2 |
130 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000376445.3 |
- |
2 |
120 |
|
Chr. X - 48973723 - 49002264 [-] |
View Full Product Details