Gene Transcripts
- Gene Symbol
- LOC102723649
- Entrez Gene ID
- 102723649
- Gene Name
- uncharacterized LOC102723649
- Gene Aliases
- Chromosome Location
- Chr. 6: 137665646 - 137674554 on Build GRCh38
- UniGene ID
- Hs.129235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125867.1 |
|
4 |
431 |
|
GenBank mRNA |
BX098901.1 |
- |
4 |
446 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000440397.1 |
- |
3 |
434 |
|
Chr. 6 - 137673460 - 137674554 [+] |
View Full Product Details