Gene Transcripts
- Gene Symbol
- LOC102724511
- Entrez Gene ID
- 102724511
- Gene Name
- uncharacterized LOC102724511
- Gene Aliases
- Chromosome Location
- Chr. 6: 170160642 - 170163027 on Build GRCh38
- UniGene ID
- Hs.550045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125878.1 |
|
3 |
509 |
|
GenBank mRNA |
BX092407.1 |
- |
3 |
479 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000429305.1 |
- |
2 |
408 |
|
Chr. 6 - 170162517 - 170163027 [+] |
View Full Product Details