Gene Transcripts
- Gene Symbol
- UBOX5-AS1
- Entrez Gene ID
- 100134015
- Gene Name
- UBOX5 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 3106911 - 3150867 on Build GRCh38
- UniGene ID
- Hs.129448
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038395.1 |
|
4 |
1146 |
|
GenBank mRNA |
AK097516.1 |
- |
4 |
1146 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000446537.5 |
- |
4 |
1144 |
|
Chr. 20 - 3106913 - 3150867 [+] |
View Full Product Details