Gene Transcripts
- Gene Symbol
- ANKAR
- Entrez Gene ID
- 150709
- Gene Name
- ankyrin and armadillo repeat containing
- Gene Aliases
- Chromosome Location
- Chr. 2: 189674292 - 189763059 on Build GRCh38
- UniGene ID
- Hs.60772
- Species
- Homo sapiens
- Gene Symbol
- OSGEPL1-AS1
- Entrez Gene ID
- 101409258
- Gene Name
- OSGEPL1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 189762780 - 189765556 on Build GRCh38
- UniGene ID
- Hs.738558
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000520651.1 |
- |
1 |
244 |
|
Chr. 2 - 189762704 - 189765556 [+] |
View Full Product Details