Gene Transcripts
- Gene Symbol
- LOC284395
- Entrez Gene ID
- 284395
- Gene Name
- uncharacterized LOC284395
- Gene Aliases
- Chromosome Location
- Chr. 19: 29287011 - 29525752 on Build GRCh38
- UniGene ID
- Hs.631601
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040029.1 |
|
10 |
2072 |
|
GenBank mRNA |
AK094793.1 |
- |
10 |
2075 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000582581.5 |
- |
10 |
2072 |
|
Chr. 19 - 29287011 - 29525752 [-] |
View Full Product Details