Gene Transcripts
- Gene Symbol
- Gm38483
- Entrez Gene ID
- 102636037
- Gene Name
- predicted gene, 38483
- Gene Aliases
- Chromosome Location
- Chr. 11: 100689745 - 100697529 on Build GRCm38
- UniGene ID
- Mm.343137
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053109.1 |
- |
|
233 |
|
View Full Product Details