Gene Transcripts
- Gene Symbol
- Gm16336
- Entrez Gene ID
- 100504184
- Gene Name
- predicted gene 16336
- Gene Aliases
- OTTMUSG00000033852
- Chromosome Location
- Chr. 7: 110904985 - 110905851 on Build GRCm38
- UniGene ID
- Mm.441327
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045267.1 |
|
3 |
356 |
|
GenBank mRNA |
BE633821.1 |
- |
3 |
356 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX517746.1 |
- |
3 |
357 |
|
Chr. 7 - 110904985 - 110905851 [+] |
View Full Product Details