Gene Transcripts
- Gene Symbol
- LOC105371730
- Entrez Gene ID
- 105371730
- Gene Name
- uncharacterized LOC105371730
- Gene Aliases
- Chromosome Location
- Chr. 17: 32141226 - 32143135 on Build GRCh38
- UniGene ID
- Hs.673781
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136413.1 |
|
2 |
1257 |
|
View Full Product Details