Gene Transcripts

Gene Symbol
Slc39a14
Entrez Gene ID
213053
Gene Name
solute carrier family 39 (zinc transporter), member 14
Gene Aliases
FAD-123, ZIP-14, Zip14, fad123
Chromosome Location
Chr. 14: 70303467 - 70351459 on Build GRCm38
UniGene ID
Mm.270647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135151.1 NP_001128623.1 6, 7 1010
NM_001135152.1 NP_001128624.1 6, 7 976
NM_144808.4 NP_659057.2 6, 7 1010
GenBank mRNA AB177995.1 - 6, 7 1013
AK090193.1 - 6, 7 973
AK192726.1 - 1, 2 77
BC021530.1 - 6, 7 1013 5353717
CB521415.1 - 3, 4 338

View Full Product Details