Gene Transcripts

Gene Symbol
Trmt2b
Entrez Gene ID
215201
Gene Name
TRM2 tRNA methyltransferase 2B
Gene Aliases
4732479N06Rik
Chromosome Location
Chr. X: 134222955 - 134277039 on Build GRCm38
UniGene ID
Mm.289368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167994.1 NP_001161466.1 3 373
NM_001167995.1 NP_001161467.1 2 297
NM_172540.2 NP_766128.2 2 297
GenBank mRNA AK028999.1 - 2 253
AK046928.1 - 2 297
AK049523.1 - 2 295
AK129030.1 - 3 316
AK137595.1 - 259
AK188477.1 - 2 190
BC056937.1 - 3 314 5716144

View Full Product Details