Gene Transcripts

Gene Symbol
Gm4846
Entrez Gene ID
226601
Gene Name
predicted gene 4846
Gene Aliases
EG226601, Fmo13
Chromosome Location
Chr. 1: 166482599 - 166506135 on Build GRCm38
UniGene ID
Mm.475593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164306.1 NP_001157778.1 1, 2 211
GenBank mRNA BC147800.1 - 1, 2 211
BC147804.1 - 1, 2 219 9087521

View Full Product Details