Gene Transcripts

Gene Symbol
Slc17a9
Entrez Gene ID
228993
Gene Name
solute carrier family 17, member 9
Gene Aliases
1700019H03Rik, BC019537, Vnut
Chromosome Location
Chr. 2: 180725213 - 180742280 on Build GRCm38
UniGene ID
Mm.87189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183161.3 NP_898984.3 3, 4 452
GenBank mRNA BC019537.1 - 2, 3 767
BG087579.2 - 3, 4 452

View Full Product Details