Gene Transcripts
- Gene Symbol
- Ap5z1
- Entrez Gene ID
- 231855
- Gene Name
- adaptor-related protein complex 5, zeta 1 subunit
- Gene Aliases
- C330006K01Rik
- Chromosome Location
- Chr. 5: 142463205 - 142478716 on Build GRCm38
- UniGene ID
- Mm.78250
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172725.2 |
NP_766313.2
|
7, 8 |
1084 |
|
GenBank mRNA |
AK040924.1 |
- |
5, 6 |
636 |
|
AK129135.1 |
- |
5, 6 |
676 |
|
AK146431.1 |
- |
7, 8 |
962 |
|
AK152402.1 |
- |
7, 8 |
962 |
|
AK153787.1 |
- |
7, 8 |
957 |
|
BC050232.1 |
- |
5, 6 |
681 |
|
BC057548.1 |
- |
7, 8 |
939 |
|
BC082786.1 |
- |
7, 8 |
1057 |
|
BC099419.1 |
- |
7, 8 |
1016 |
|
BC144755.1 |
- |
5, 6 |
615 |
|
View Full Product Details