Gene Transcripts

Gene Symbol
Itga8
Entrez Gene ID
241226
Gene Name
integrin alpha 8
Gene Aliases
AI447669
Chromosome Location
Chr. 2: 12106632 - 12301996 on Build GRCm38
UniGene ID
Mm.329997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001309.2 NP_001001309.1 20 2199
GenBank mRNA AF041409.1 - 20 1930
AK031326.1 - 5 505
BC137793.1 - 21 2184

View Full Product Details