Gene Transcripts
- Gene Symbol
- Olfr788
- Entrez Gene ID
- 258544
- Gene Name
- olfactory receptor 788
- Gene Aliases
- MOR111-4
- Chromosome Location
- Chr. 10: 129472694 - 129473629 on Build GRCm38
- UniGene ID
- Mm.377560
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146551.1 |
NP_666762.1
|
1 |
497 |
|
View Full Product Details