Gene Transcripts

Gene Symbol
Olfr1161
Entrez Gene ID
258845
Gene Name
olfactory receptor 1161
Gene Aliases
MOR174-2
Chromosome Location
Chr. 2: 88024702 - 88025713 on Build GRCm38
UniGene ID
Mm.377718
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146848.2 NP_667059.2 1 603
GenBank mRNA BC131999.1 - 1 603
BC132001.1 - 1 603 40130278

View Full Product Details