Gene Transcripts

Gene Symbol
Olfr644
Entrez Gene ID
259125
Gene Name
olfactory receptor 644
Gene Aliases
MOR13-1
Chromosome Location
Chr. 7: 104068085 - 104069029 on Build GRCm38
UniGene ID
Mm.377862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147121.1 NP_667332.1 1 416
GenBank mRNA BC099887.1 - 1 408

View Full Product Details