Gene Transcripts
- Gene Symbol
- A830018L16Rik
- Entrez Gene ID
- 320492
- Gene Name
- RIKEN cDNA A830018L16 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 11414105 - 11975902 on Build GRCm38
- UniGene ID
- Mm.52021
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160369.2 |
NP_001153841.1
|
1, 2 |
542 |
|
NM_001160370.2 |
NP_001153842.1
|
1, 2 |
542 |
|
NM_001160371.2 |
NP_001153843.1
|
1, 2 |
542 |
|
NM_177173.6 |
NP_796147.2
|
1, 2 |
542 |
|
GenBank mRNA |
AK034391.1 |
- |
1, 2 |
538 |
|
AK039555.1 |
- |
1, 2 |
349 |
|
AK043673.1 |
- |
1, 2 |
379 |
|
AK081622.1 |
- |
1, 2 |
543 |
|
AK135047.1 |
- |
1, 2 |
536 |
|
AK136763.1 |
- |
1, 2 |
343 |
|
BB649073.1 |
- |
1, 2 |
543 |
|
BC137903.1 |
- |
1, 2 |
138 |
8860922 |
BC145022.1 |
- |
1, 2 |
138 |
9053570 |
BC145023.1 |
- |
1, 2 |
138 |
9053571 |
View Full Product Details