Gene Transcripts

Gene Symbol
Tigd3
Entrez Gene ID
332359
Gene Name
tigger transposable element derived 3
Gene Aliases
Chromosome Location
Chr. 19: 5891138 - 5895166 on Build GRCm38
UniGene ID
Mm.329642
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198634.1 NP_941036.1 2 300
GenBank mRNA BC053500.2 - 2 339
BC055784.1 - 2 300 6404652

View Full Product Details