Gene Transcripts
- Gene Symbol
- 9930105H17Rik
- Entrez Gene ID
- 100669
- Gene Name
- RIKEN cDNA 9930105H17 gene
- Gene Aliases
- AI462335
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.296839
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037062.1 |
- |
1 |
1429 |
|
BC019681.1 |
- |
3 |
1835 |
|
BC027236.1 |
- |
1 |
840 |
|
- Gene Symbol
- Gm8113
- Entrez Gene ID
- 666451
- Gene Name
- predicted gene 8113
- Gene Aliases
- EG666451
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.390434
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142214.1 |
- |
|
1683 |
|
View Full Product Details