Gene Transcripts
- Gene Symbol
- Tsen15
- Entrez Gene ID
- 66637
- Gene Name
- tRNA splicing endonuclease 15 homolog (S. cerevisiae)
- Gene Aliases
- 5730449L18Rik, AL023077, Sen15
- Chromosome Location
- Chr. 1: 152370735 - 152386682 on Build GRCm38
- UniGene ID
- Mm.379120
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC024342.1 |
- |
1, 2 |
154 |
|
BC145701.1 |
- |
1, 2 |
191 |
40131004 |
View Full Product Details