Gene Transcripts

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
Chromosome Location
Chr. 11: 67428460 - 67435408 on Build GRCh38
UniGene ID
Hs.534345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003952.2 NP_003943.2 12 1066
XM_005274164.1 XP_005274221.1 11 1014
XM_005274165.4 XP_005274222.1 11 891
XM_006718655.3 XP_006718718.1 13 1187
XM_006718656.3 XP_006718719.1 12 1135
XM_006718657.1 XP_006718720.1 12 1120
XM_017018108.1 XP_016873597.1 13 1198
GenBank mRNA AB016869.1 - 12 1038
AB019245.1 - 12 1033
AB208827.1 - 6 4539
AF076931.1 - 12 1019
AF099739.1 - 12 1023
BC000094.2 - 12 1039 3508140
BC006106.1 - 12 1022 2959036
BC136564.1 - 12 1040 9020554
BX538119.1 - 4 221

View Full Product Details