Gene Transcripts

Gene Symbol
Cd300lh
Entrez Gene ID
382551
Gene Name
CD300 antigen like family member H
Gene Aliases
AY457049, Clm3, Lmir7, clm-3
Chromosome Location
Chr. 11: 115040520 - 115048419 on Build GRCm38
UniGene ID
Mm.424732
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199201.1 NP_954671.1 4 683
GenBank mRNA AY457049.1 - 4 683
BC119352.1 - 4 718 8734101
BC120623.1 - 718 40129546

Gene Symbol
Gm11710
Entrez Gene ID
100043123
Gene Name
predicted gene 11710
Gene Aliases
OTTMUSG00000003605
Chromosome Location
Chr. 11: 115020689 - 115027026 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101656.2 NP_001095126.1 718

Gene Symbol
Gm11711
Entrez Gene ID
100043125
Gene Name
predicted gene 11711
Gene Aliases
OTTMUSG00000003606
Chromosome Location
Chr. 11: 115031447 - 115037780 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101657.2 NP_001095127.1 4 718

View Full Product Details