Gene Transcripts

Gene Symbol
Sct
Entrez Gene ID
24769
Gene Name
secretin
Gene Aliases
Secr
Chromosome Location
Chr. 1: 214264865 - 214270337 on Build Rnor_6.0
UniGene ID
Rn.9919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022670.2 NP_073161.1 3 264
XM_006230518.2 XP_006230580.1 4 512
GenBank mRNA M31495.1 - 3 287

View Full Product Details