Gene Transcripts

Gene Symbol
Sult2a6
Entrez Gene ID
24902
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
Gene Aliases
RATSMP2A, RATSMP2B, Smp2a, St2a2, Sult2a4l1, Sult2al1
Chromosome Location
Chr. 1: 76529121 - 76722965 on Build Rnor_6.0
UniGene ID
Rn.221887
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012695.3 NP_036827.3 3 482
GenBank mRNA FQ209484.1 - 3 357
FQ209563.1 - 3 357
FQ218388.1 - 3 357
FQ218712.1 - 3 357
FQ219267.1 - 3 357
FQ219345.1 - 3 357
M33329.1 - 3 482
X63410.1 - 3 482

View Full Product Details