Gene Transcripts

Gene Symbol
P3h4
Entrez Gene ID
59101
Gene Name
prolyl 3-hydroxylase family member 4
Gene Aliases
Leprel4, Sc65
Chromosome Location
Chr. 10: 88318231 - 88325524 on Build Rnor_6.0
UniGene ID
Rn.90100
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021581.1 NP_067592.1 8 1271
XM_006247414.2 XP_006247476.1 8 1576
GenBank mRNA FQ227296.1 - 8 1378
X65454.1 - 8 1271

View Full Product Details