Gene Transcripts
- Gene Symbol
- SP3
- Entrez Gene ID
- 6670
- Gene Name
- Sp3 transcription factor
- Gene Aliases
- SPR2
- Chromosome Location
- Chr. 2: 173906459 - 173965702 on Build GRCh38
- UniGene ID
- Hs.531587
- Species
- Homo sapiens
- Gene Symbol
- SP3P
- Entrez Gene ID
- 160824
- Gene Name
- Sp3 transcription factor pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 13: 89359753 - 89363502 on Build GRCh38
- UniGene ID
- Hs.407509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036697.1 |
- |
1 |
1417 |
|
View Full Product Details