Gene Transcripts

Gene Symbol
Prkd1
Entrez Gene ID
85421
Gene Name
protein kinase D1
Gene Aliases
Pkcm, Prkcm, nPKC-D1, nPKC-mu
Chromosome Location
Chr. 6: 71035017 - 71349531 on Build Rnor_6.0
UniGene ID
Rn.144619
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276715.1 NP_001263644.1 8 1061
XM_006240074.2 XP_006240136.1 7 1319

View Full Product Details