Gene Transcripts

Gene Symbol
C9
Entrez Gene ID
117512
Gene Name
complement component 9
Gene Aliases
Chromosome Location
Chr. 2: 55775562 - 55823807 on Build Rnor_6.0
UniGene ID
Rn.10252
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057146.1 NP_476487.1 6 784
GenBank mRNA BC091127.1 - 6 780
U49071.1 - 6 766
U52948.1 - 6 784

View Full Product Details