Gene Transcripts

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
Chromosome Location
Chr. 9: 27109141 - 27230178 on Build GRCh38
UniGene ID
Hs.89640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000459.4 NP_000450.2 10 1899
NM_001290077.1 NP_001277006.1 9 1770
NM_001290078.1 NP_001277007.1 8 1458
XM_005251561.1 XP_005251618.1 10 1566
XM_005251563.1 XP_005251620.1 9 1437
GenBank mRNA AB086825.1 - 9 1426
AB208796.1 - 10 1577
AK291775.1 - 10 1587
AK294887.1 - 8 1459
AK295043.1 - 9 1496
AK308447.1 - 7 1010
BC035514.1 - 10 1574 5228999
EU826592.1 - 9 1320
L06139.1 - 10 1597

View Full Product Details