Gene Transcripts

Gene Symbol
Flii
Entrez Gene ID
287375
Gene Name
flightless I homolog (Drosophila)
Gene Aliases
Fliih
Chromosome Location
Chr. 10: 46955460 - 46969457 on Build Rnor_6.0
UniGene ID
Rn.144698
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008279.1 NP_001008280.1 8 791
XM_006246468.2 XP_006246530.1 8 842
GenBank mRNA BC085829.1 - 8 791

View Full Product Details