Gene Transcripts

Gene Symbol
St6galnac1
Entrez Gene ID
287920
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
Siat7A
Chromosome Location
Chr. 10: 105713305 - 105723700 on Build Rnor_6.0
UniGene ID
Rn.198923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105859.1 NP_001099329.1 7 1373
GenBank mRNA AJ634458.1 - 7 1272

View Full Product Details