Gene Transcripts

Gene Symbol
Ifnar1
Entrez Gene ID
288264
Gene Name
interferon (alpha, beta and omega) receptor 1
Gene Aliases
Chromosome Location
Chr. 11: 31640407 - 31664067 on Build Rnor_6.0
UniGene ID
Rn.105738
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105893.1 NP_001099363.2 6, 7 1062
NM_001271316.1 NP_001258245.1 7, 8 1252
GenBank mRNA FQ210936.1 - 7, 8 1052

View Full Product Details