Gene Transcripts

Gene Symbol
Zfp473
Entrez Gene ID
292884
Gene Name
zinc finger protein 473
Gene Aliases
RGD1305084, Znf473
Chromosome Location
Chr. 1: 100736319 - 100755496 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229185.1 XP_006229247.1 5 1261
XM_006229186.1 XP_006229248.1 5 879
XM_006229187.1 XP_006229249.1 5 832
XM_006229188.2 XP_006229250.1 5 760
XM_008759453.1 XP_008757675.1 4 1084
XM_008759454.1 XP_008757676.1 4 628

View Full Product Details