Gene Transcripts

Gene Symbol
Vom2r41
Entrez Gene ID
293069
Gene Name
vomeronasal 2 receptor, 41
Gene Aliases
RGD1561322
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.217629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099500.1 NP_001092970.1 3 1086

View Full Product Details