Gene Transcripts

Gene Symbol
LOC689730
Entrez Gene ID
689730
Gene Name
similar to ubiquitin specific peptidase 17 (homolog)
Gene Aliases
Chromosome Location
Chr. Un|NW_007906785: 221269 - 224025 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773797.1 XP_008772019.1 576

Gene Symbol
LOC100910107
Entrez Gene ID
100910107
Gene Name
ubiquitin carboxyl-terminal hydrolase DUB-1-like
Gene Aliases
Chromosome Location
Chr. 1: 169720247 - 169721788 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759844.1 XP_008758066.1 1 518

View Full Product Details