Gene Transcripts

Gene Symbol
Ndufaf5
Entrez Gene ID
296190
Gene Name
NADH dehydrogenase (ubiquinone) complex I, assembly factor 5
Gene Aliases
RGD1309829
Chromosome Location
Chr. 3: 133232408 - 133261932 on Build Rnor_6.0
UniGene ID
Rn.98718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126371.1 NP_001119843.1 7, 8 750
GenBank mRNA BC166551.1 - 7, 8 750
FQ213977.1 - 7, 8 716

View Full Product Details