Gene Transcripts

Gene Symbol
Cst7
Entrez Gene ID
296257
Gene Name
cystatin F (leukocystatin)
Gene Aliases
Chromosome Location
Chr. 3: 146363915 - 146374297 on Build Rnor_6.0
UniGene ID
Rn.24282
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235213.2 XP_006235275.1 2, 3 1491
XM_008762305.1 XP_008760527.1 2, 3 1491
XM_008762306.1 XP_008760528.1 2, 3 1488

View Full Product Details