Gene Transcripts

Gene Symbol
Tssk3
Entrez Gene ID
297891
Gene Name
testis-specific serine kinase 3
Gene Aliases
Stk22c, Tssk-3
Chromosome Location
Chr. 5: 147689090 - 147691930 on Build Rnor_6.0
UniGene ID
Rn.109479
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007650.1 NP_001007651.1 2 387
XM_006238925.2 XP_006238987.1 2 162
GenBank mRNA AY346102.1 - 2 387

View Full Product Details