Gene Transcripts

Gene Symbol
Etfa
Entrez Gene ID
300726
Gene Name
electron-transfer-flavoprotein, alpha polypeptide
Gene Aliases
ETF
Chromosome Location
Chr. 8: 60028786 - 60086352 on Build Rnor_6.0
UniGene ID
Rn.32496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009668.1 NP_001009668.1 2 148
GenBank mRNA BC088412.1 - 2 148
FQ219339.1 - 2 171

View Full Product Details