Gene Transcripts
- Gene Symbol
- Arsg
- Entrez Gene ID
- 303631
- Gene Name
- arylsulfatase G
- Gene Aliases
- RGD1306571
- Chromosome Location
- Chr. 10: 97722550 - 97859975 on Build Rnor_6.0
- UniGene ID
- Rn.221856
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001047877.1 |
NP_001041342.1
|
1, 2 |
220 |
|
XM_006247599.2 |
XP_006247661.1
|
3, 4 |
2420 |
|
XM_006247600.2 |
XP_006247662.1
|
2, 3 |
2277 |
|
XM_006247601.2 |
XP_006247663.1
|
4, 5 |
2646 |
|
XM_006247602.2 |
XP_006247664.1
|
3, 4 |
1208 |
|
XM_006247603.2 |
XP_006247665.1
|
3, 4 |
1062 |
|
XM_006247604.2 |
XP_006247666.1
|
3, 4 |
2478 |
|
XM_006247606.2 |
XP_006247668.1
|
2, 3 |
2335 |
|
XM_006247607.2 |
XP_006247669.1
|
2, 3 |
1247 |
|
XM_006247608.2 |
XP_006247670.1
|
1, 2 |
897 |
|
XM_008768368.1 |
XP_008766590.1
|
4, 5 |
2589 |
|
XM_008768369.1 |
XP_008766591.1
|
5, 6 |
2053 |
|
View Full Product Details